Learn More
Alkali Scientific Acrylamide, 40% Solution, Liquid, 1 L
SDP
Catalog No. 5000660965
Encompass_Preferred
Acrylamide 40% solution (1L) is a ready-to-use liquid reagent designed for electrophoresis and gel preparation. This highly concentrated solution simplifies the process of creating polyacrylamide gels, as it eliminates the need for measuring and dissolving solid acrylamide. It is commonly used in protein and nucleic acid electrophoresis to separate biomolecules based on their charge and size. The solution’s high concentration allows for easy dilution, making it ideal for laboratory use in gel preparation, biochemical assays, and other molecular biology techniques. Proper handling and storage are critical due to acrylamide's potential neurotoxic properties, requiring adherence to safety guidelines.
Catalog No. 50-006-60965 Supplier Alkali Scientific Supplier No. ACR006CP
Acrylamide 40% solution (1L) is a ready-to-use liquid reagent designed for electrophoresis and gel preparation. This highly concentrated solution simplifies the process of creating polyacrylamide gels, as it eliminates the need for measuring and dissolving solid acrylamide. It is commonly used in protein and nucleic acid electrophoresis to separate biomolecules based on their charge and size. The solution’s high concentration allows for easy dilution, making it ideal for laboratory use in gel preparation, biochemical assays, and other molecular biology techniques. Proper handling and storage are critical due to acrylamide's potential neurotoxic properties, requiring adherence to safety guidelines.