Learn More
Crescent Chemical Co Inc ACRYLAMIDE 4X MOL. BIOL. GRADE
SDP
Catalog No. 50247128
Encompass_Preferred
Acrylamide 4X molecular biology grade DNase and RNase: non-detectable. Special quality for use in molecular biological applications as well as all electrophoresis techniques. Size - 100G Storage Conditions - +2 °C TO +8 °C Catalog Number - 10678.0
Catalog No. 50-247-128 Supplier Crescent Chemical Co Inc Supplier No. SE10678.02
Acrylamide 4X molecular biology grade DNase and RNase: non-detectable. Special quality for use in molecular biological applications as well as all electrophoresis techniques. Size - 100G Storage Conditions - +2 °C TO +8 °C Catalog Number - 10678.0