Learn More
Crescent Chemical Co Inc ACRYLAMIDE 4X SOLUTION (40%)
SDP
Catalog No. 50247127
Encompass_Preferred
Acrylamide 4X solution (40 % w/v) Solution contains 40 % (w/v) highly purified acrylamide in deionized water. Size - 1L Storage Conditions - +2 °C TO +8 °C Catalog Number - 10677.01
Catalog No. 50-247-127 Supplier Crescent Chemical Co Inc Supplier No. SE10677.01
Acrylamide 4X solution (40 % w/v) Solution contains 40 % (w/v) highly purified acrylamide in deionized water. Size - 1L Storage Conditions - +2 °C TO +8 °C Catalog Number - 10677.01