Learn More
Research Products International Corp Acrylamide, 500 G
SDP
Catalog No. 501977598
Encompass_Preferred
Acrylamide (C₃H₅NO) is a water-soluble monomer (71.08 g/mol) that forms polyacrylamide gels used in electrophoresis to separate proteins and nucleic acids. It polymerizes with bis-acrylamide and initiators like APS and TEMED. Common applications in life science labs include SDS-PAGE, Western blotting, and nucleic acid analysis.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7598 Supplier Research Products International Corp Supplier No. A11260500.0
Acrylamide (C₃H₅NO) is a water-soluble monomer (71.08 g/mol) that forms polyacrylamide gels used in electrophoresis to separate proteins and nucleic acids. It polymerizes with bis-acrylamide and initiators like APS and TEMED. Common applications in life science labs include SDS-PAGE, Western blotting, and nucleic acid analysis.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.