Learn More
Acrylamide-bis-acryl, 40% (w/v), 29:1 Solution, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsDescription
- Acrylamide/bis-acrylamide (40% w/v) 29:1 Solution is convenient ready-to-use solutions of 4X crystalized high quality acrylamide for molecular biology, and NN'-Methylenebisacrylamide in deionised water.
- Premixed 19:1 liquid solution eliminates the need to weigh toxic acrylamide and bis-acrylamide.
- The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
- Acrylamide/bis-acrylamide (40% w/v) 29:1 Solution is prepared from highly purified acrylamide (4X) and bis-acrylamide (2X) using deionized ultrafiltered water.
Specifications
Specifications
| Absorbance | A290 (1% in water): ≤0.1 |
| Physical Form | Liquid |
| Composition | Acrylamide/Bis-acrylamide 29:1 |
| Quantity | 1 L |
| Grade | Molecular Biology |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.