Learn More
Research Products International Corp Acrylamide/bis-Acrylamide, 29:1, 5 L
SDP
Catalog No. 501977033
Encompass_Preferred
A 40% solution containing 38.67% (w/v) Acrylamide and 1.33% (w/v) bis-Acrylamide for a monomer to crosslinker ratio of 29:1.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7033 Supplier Research Products International Corp Supplier No. A114055000.0
A 40% solution containing 38.67% (w/v) Acrylamide and 1.33% (w/v) bis-Acrylamide for a monomer to crosslinker ratio of 29:1.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.