Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 19:1 Ratio Solution, 500 Milliliters
SDP
Catalog No. 5000660722
Encompass_Preferred
Acrylamide/bis Acrylamide, 19:1 Ratio Solution, 500 Milliliters, is a liquid solution used for preparing polyacrylamide gels for electrophoresis. The 19:1 ratio provides a medium that is ideal for separating larger biomolecules such as proteins, ensuring precise resolution and reliable performance. This solution eliminates the need for mixing dry acrylamide and bisacrylamide, reducing preparation time and minimizing human error. It is commonly used in protein electrophoresis, where it offers consistent results for protein separation, molecular biology research, and various analytical applications in laboratories focusing on protein studies.
Catalog No. 50-006-60722 Supplier Alkali Scientific Supplier No. CRY004CP
Acrylamide/bis Acrylamide, 19:1 Ratio Solution, 500 Milliliters, is a liquid solution used for preparing polyacrylamide gels for electrophoresis. The 19:1 ratio provides a medium that is ideal for separating larger biomolecules such as proteins, ensuring precise resolution and reliable performance. This solution eliminates the need for mixing dry acrylamide and bisacrylamide, reducing preparation time and minimizing human error. It is commonly used in protein electrophoresis, where it offers consistent results for protein separation, molecular biology research, and various analytical applications in laboratories focusing on protein studies.