Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 19:1 Ratio Solution, Liquid, 1 L
SDP
Catalog No. 5000660835
Encompass_Preferred
Acrylamide/bis Acrylamide, 19:1 Ratio Solution, Liquid, 1 L, is a ready-to-use solution ideal for polyacrylamide gel electrophoresis (PAGE). The 19:1 ratio ensures optimal gel formation for separating larger proteins and biomolecules. This solution is pre-mixed, saving time and reducing the need for precise measurements during gel preparation. It is widely used in molecular biology, biochemistry, and protein analysis to achieve high-resolution separation of proteins based on size. The solution is convenient for researchers who require reliable and reproducible results in electrophoresis experiments, particularly in proteomics and enzymology studies.
Catalog No. 50-006-60835 Supplier Alkali Scientific Supplier No. CRY003CP
Acrylamide/bis Acrylamide, 19:1 Ratio Solution, Liquid, 1 L, is a ready-to-use solution ideal for polyacrylamide gel electrophoresis (PAGE). The 19:1 ratio ensures optimal gel formation for separating larger proteins and biomolecules. This solution is pre-mixed, saving time and reducing the need for precise measurements during gel preparation. It is widely used in molecular biology, biochemistry, and protein analysis to achieve high-resolution separation of proteins based on size. The solution is convenient for researchers who require reliable and reproducible results in electrophoresis experiments, particularly in proteomics and enzymology studies.