Learn More
Sigma Aldrich Fine Chemicals Biosciences Acrylamide/Bis-acrylamide
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A35745X100ML
Encompass
Polyacrylamide gels are prepared using acrylamide and bis-acrylamide where bis-acrylamide is a cross-linking agent. On the other hand acrylamide is a water soluble monomer which helps in forming a transparent stable insoluble gel.
Catalog No. 50-290-0350