Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 29:1 Ratio Solution, 500 Milliliters
SDP
Catalog No. 5000660771
Encompass_Preferred
Acrylamide/bis Acrylamide, 29:1 Ratio Solution, 500 Milliliters, is a liquid solution optimized for polyacrylamide gel electrophoresis (PAGE) to separate small proteins and nucleic acids. The 29:1 ratio ensures the production of highly porous gels, which enhances the separation efficiency of smaller molecules. This pre-mixed solution is ready for use, eliminating the need for individual measurements and reducing preparation time. It is widely used in research labs, particularly for protein analysis, RNA separation, and various molecular biology applications where precise separation and resolution of small biomolecules are required.
Catalog No. 50-006-60771 Supplier Alkali Scientific Supplier No. CRY007CP
Acrylamide/bis Acrylamide, 29:1 Ratio Solution, 500 Milliliters, is a liquid solution optimized for polyacrylamide gel electrophoresis (PAGE) to separate small proteins and nucleic acids. The 29:1 ratio ensures the production of highly porous gels, which enhances the separation efficiency of smaller molecules. This pre-mixed solution is ready for use, eliminating the need for individual measurements and reducing preparation time. It is widely used in research labs, particularly for protein analysis, RNA separation, and various molecular biology applications where precise separation and resolution of small biomolecules are required.