Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 29:1 Ratio Solution, Liquid, 1 L
SDP
Catalog No. 5000660686
Encompass_Preferred
Acrylamide/bis Acrylamide, 29:1 Ratio Solution, Liquid, 1 L, is a ready-to-use solution designed for high-resolution electrophoresis. The 29:1 ratio of acrylamide to bis-acrylamide creates a more porous gel matrix, ideal for separating smaller proteins and nucleic acids in polyacrylamide gel electrophoresis (PAGE). This liquid solution eliminates the need for manual mixing of dry reagents, saving time and reducing the potential for error. It is particularly suited for applications that require precise separation of small biomolecules, such as in proteomics, nucleic acid research, and molecular biology studies.
Catalog No. 50-006-60686 Supplier Alkali Scientific Supplier No. CRY006CP
Acrylamide/bis Acrylamide, 29:1 Ratio Solution, Liquid, 1 L, is a ready-to-use solution designed for high-resolution electrophoresis. The 29:1 ratio of acrylamide to bis-acrylamide creates a more porous gel matrix, ideal for separating smaller proteins and nucleic acids in polyacrylamide gel electrophoresis (PAGE). This liquid solution eliminates the need for manual mixing of dry reagents, saving time and reducing the potential for error. It is particularly suited for applications that require precise separation of small biomolecules, such as in proteomics, nucleic acid research, and molecular biology studies.