Learn More
Sigma Aldrich Fine Chemicals Biosciences Acrylamide/Bis acrylamide
Catalog No. 502905456
Encompass
Polyacrylamide gels are prepared using acrylamide and bis-acrylamide where bis-acrylamide is a cross-linking agent. On the other hand acrylamide is a water soluble monomer which helps in forming a transparent stable insoluble gel.
Catalog No. 50-290-5456 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A35745L
Polyacrylamide gels are prepared using acrylamide and bis-acrylamide where bis-acrylamide is a cross-linking agent. On the other hand acrylamide is a water soluble monomer which helps in forming a transparent stable insoluble gel.