Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylamide/Bis-acrylamide 37, 5:1, MP Biomedicals™

Catalog No. MP1BIAC3752 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1BIAC3752 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1BIAC3752 Supplier MP Biomedicals, Inc Supplier No. 11BIAC3752
Add to Cart
Add to Cart

Acrylamide/Bis-acrylamide 37, 5:1, Molecular Biology Grade, 4X Crystallized, Solution

  • Acrylamide/Bis-acrylamide (40% w/v solution) is prepared from highly purified acrylamide (4X) and bis-acrylamide (2X) using deionized ultrafiltered water.
  • It is used for the separation of high molecular weight proteins.
  • It is convenient ready-to-use solutions of 4X crystalized high quality acrylamide for molecular biology, and NN'-Methylenebisacrylamide in deonised water.
  • Premixed 19:1 liquid solution eliminates the need to weigh toxic acrylamide and bis-acrylamide.
  • The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis-Acrylamide Gel
Physical Form Liquid
Quantity 1 L
Concentration or Composition (by Analyte or Components) 40% (w/v)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.