Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 37.5:1 Ratio Solution, 5 Liters
SDP
Catalog No. 5000660840
Encompass_Preferred
Acrylamide/Bis Acrylamide, 37.5:1 Ratio Solution, 5 Liters is a ready-to-use solution for polymerization applications requiring a precise and consistent 37.5:1 ratio of acrylamide to bis-acrylamide. This solution simplifies gel preparation for SDS-PAGE, protein separation, and other molecular biology techniques by eliminating the need for mixing powders. The 5-liter volume is ideal for large-scale applications in research laboratories and industrial settings where a high volume of polymerization is required. The solution is designed for use in electrophoresis and gel preparation, ensuring consistent gel formation, resolution, and reproducibility in various protein analysis workflows.
Catalog No. 50-006-60840 Supplier Alkali Scientific Supplier No. CRY017CP
Acrylamide/Bis Acrylamide, 37.5:1 Ratio Solution, 5 Liters is a ready-to-use solution for polymerization applications requiring a precise and consistent 37.5:1 ratio of acrylamide to bis-acrylamide. This solution simplifies gel preparation for SDS-PAGE, protein separation, and other molecular biology techniques by eliminating the need for mixing powders. The 5-liter volume is ideal for large-scale applications in research laboratories and industrial settings where a high volume of polymerization is required. The solution is designed for use in electrophoresis and gel preparation, ensuring consistent gel formation, resolution, and reproducibility in various protein analysis workflows.