Learn More
Alkali Scientific Acrylamide/bis Acrylamide, 37.5:1 Ratio Solution, 500 Milliliters
SDP
Catalog No. 5000660832
Encompass_Preferred
Acrylamide/bis Acrylamide, 37.5:1 Ratio Solution, 500 Milliliters, is a liquid solution for preparing polyacrylamide gels that separate larger proteins and other large biomolecules. The 37.5:1 acrylamide-to-bis-acrylamide ratio creates a gel matrix that is ideal for resolving larger molecules in electrophoresis. This ready-to-use solution streamlines the gel preparation process by eliminating the need for mixing dry reagents. It is commonly used in protein research, biomolecule characterization, and molecular biology applications where the separation of larger proteins or nucleic acids is required for detailed analysis.
Catalog No. 50-006-60832 Supplier Alkali Scientific Supplier No. CRY010CP
Acrylamide/bis Acrylamide, 37.5:1 Ratio Solution, 500 Milliliters, is a liquid solution for preparing polyacrylamide gels that separate larger proteins and other large biomolecules. The 37.5:1 acrylamide-to-bis-acrylamide ratio creates a gel matrix that is ideal for resolving larger molecules in electrophoresis. This ready-to-use solution streamlines the gel preparation process by eliminating the need for mixing dry reagents. It is commonly used in protein research, biomolecule characterization, and molecular biology applications where the separation of larger proteins or nucleic acids is required for detailed analysis.