Learn More
Alkali Scientific Acrylamide/bis Acrylamide, Premixed Powder 19:1 Ratio, 500 G
SDP
Catalog No. 5000660690
Encompass_Preferred
Acrylamide/Bis Acrylamide, Premixed Powder, 19:1 Ratio, 500 G is a reagent used in polymerization and gel preparation for electrophoresis. This premixed powder offers a consistent ratio of acrylamide to bis-acrylamide, allowing precise control over the crosslinking and polymerization process. The 19:1 ratio is ideal for preparing gels for SDS-PAGE (Sodium Dodecyl Sulfate Polyacrylamide Gel Electrophoresis), commonly used in protein analysis. The product ensures high-quality gels with good resolution and reproducibility, essential for molecular biology and protein chemistry experiments. This reagent is widely used in laboratories for protein separation, characterization, and Western blotting applications.
Catalog No. 50-006-60690 Supplier Alkali Scientific Supplier No. CRY012CP
Acrylamide/Bis Acrylamide, Premixed Powder, 19:1 Ratio, 500 G is a reagent used in polymerization and gel preparation for electrophoresis. This premixed powder offers a consistent ratio of acrylamide to bis-acrylamide, allowing precise control over the crosslinking and polymerization process. The 19:1 ratio is ideal for preparing gels for SDS-PAGE (Sodium Dodecyl Sulfate Polyacrylamide Gel Electrophoresis), commonly used in protein analysis. The product ensures high-quality gels with good resolution and reproducibility, essential for molecular biology and protein chemistry experiments. This reagent is widely used in laboratories for protein separation, characterization, and Western blotting applications.