Learn More
Alkali Scientific Acrylamide/bis Acrylamide, Premixed Powder, 50:1 Ratio, 100 G
SDP
Catalog No. 5000660259
Encompass_Preferred
Acrylamide/Bis Acrylamide, Premixed Powder, 50:1 Ratio, 100 G is a reagent that provides a low crosslinking density for gel preparation, useful for certain molecular biology and protein analysis techniques. The 50:1 ratio of acrylamide to bis-acrylamide results in gels with lower mechanical strength and higher porosity, making it ideal for applications where larger molecules need to be separated, such as RNA and large protein electrophoresis. This premixed powder ensures accurate preparation, eliminating the need for separate mixing of acrylamide and bis-acrylamide. It is used widely in electrophoresis, protein separation, and other bioanalytical applications requiring a softer, more flexible gel matrix.
Catalog No. 50-006-60259 Supplier Alkali Scientific Supplier No. CRY015CP
Acrylamide/Bis Acrylamide, Premixed Powder, 50:1 Ratio, 100 G is a reagent that provides a low crosslinking density for gel preparation, useful for certain molecular biology and protein analysis techniques. The 50:1 ratio of acrylamide to bis-acrylamide results in gels with lower mechanical strength and higher porosity, making it ideal for applications where larger molecules need to be separated, such as RNA and large protein electrophoresis. This premixed powder ensures accurate preparation, eliminating the need for separate mixing of acrylamide and bis-acrylamide. It is used widely in electrophoresis, protein separation, and other bioanalytical applications requiring a softer, more flexible gel matrix.