Learn More
Sigma Aldrich Fine Chemicals Biosciences Acrylamide/Bis acrylamide1L
Catalog No. 502905358
Encompass
Acrylamidenbsp(AA) is a neurotoxin and carcinogen. It was found in potato products like french fries and potato chips and also in several cereal-based products. This highly reactive organic compound has the ability to polymerize to form polyacrylamide.
Catalog No. 50-290-5358 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A27921L
Acrylamidenbsp(AA) is a neurotoxin and carcinogen. It was found in potato products like french fries and potato chips and also in several cereal-based products. This highly reactive organic compound has the ability to polymerize to form polyacrylamide.