Learn More
Crescent Chemical Co Inc ACRYLAMIDE/BIS SOLUTION
SDP
Supplier:  Crescent Chemical Co Inc SE10681.02
Encompass_Preferred
Acrylamide/Bis Solution, 37.5:1 (40 % w/v), 2.6 % C Solution of acrylamide and N,N'-methylene bisacrylamide (Bis) in deionized water. Convenient to use, reduced risk of neurotoxic acrylamide dust in the air. Applicable to all electrophoresis techniques. Size - 4x500ML S
Catalog No. 50-247-137