Learn More
Crescent Chemical Co Inc ACRYLAMIDE/BIS SOLUTION
SDP
Catalog No. 50247142
Encompass_Preferred
Acrylamide/Bis Solution, 37.5:1 (30 % w/v), 2.6 % C Solution of acrylamide and N,N'-methylene bisacrylamide (Bis) in deionized water. Convenient to use, reduced risk of neurotoxic acrylamide dust in the air. Applicable to all electrophoresis techniques. Size - 500ML Sto
Catalog No. 50-247-142 Supplier Crescent Chemical Co Inc Supplier No. SE10688.01
Acrylamide/Bis Solution, 37.5:1 (30 % w/v), 2.6 % C Solution of acrylamide and N,N'-methylene bisacrylamide (Bis) in deionized water. Convenient to use, reduced risk of neurotoxic acrylamide dust in the air. Applicable to all electrophoresis techniques. Size - 500ML Sto