Learn More
GenDEPOT ACRYLAMIDE:BIS40%,29:1,495ML
Catalog No. 501015464
Encompass_Preferred
Acrylamide : bis 40%, 29:1 Solution, 495ml
Catalog No. 50-101-5464 Supplier GenDEPOT Supplier No. A0421050
Acrylamide : bis 40%, 29:1 Solution, 495ml