Learn More
Medchemexpress LLC Acrylamide-d3 | 122775-19-3 | MFCD00143546 | 74.10 g/mol | C3H2D3NO | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW6537285MG
Encompass_Preferred
Acrylamide-d3 is the deuterium-labeled isotopologue of acrylamide supplied for analytical and research use as a small-scale standard. It is intended for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses; a certificate of analysis and data sheet provide analytical specifications.
  • Deuterium-labeled (d3) isotopologue of acrylamide.
  • Suitable as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Available in milligram pack sizes for research use.
  • Certificate of analysis and data sheet available for purity and handling details.
  • Molecular formula C3H2D3NO; molecular weight 74.10 g/mol.
Catalog No. 50-002-84726