Learn More
Waters Corp Acrylamide Refill Kit, LC-MS, Enhanced Clean-up, 96 Analysis
Catalog No. 502392762 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This refill kit is designed for the sample extraction and enhanced clean-up of acrylamide in processed foods using the Waters methodology. It includes consumables for an additional mixed-mode SPE clean-up step, suitable for adoption onto mid-tier UPLC-MS/MS platforms. It provides a complete sample preparation workflow for acrylamide detection in processed foods and supports enhanced solid-phase extraction clean-up. This kit is intended to replenish consumables for the Acrylamide Starter Kit.
  • Number of analysis: 96
  • Sample preparation workflow
  • One package per kit
  • UPLC separation mode
  • MS/MS detection method
  • LC-MS system type
  • Food application
  • Application kits product type
  • One unit per package
  • Refill kit for acrylamide starter kit
  • Compatibility with mid-tier UPLC-MS/MS platforms
Catalog No. 50-239-2762 Supplier Waters Corp Supplier No. 176004420
This refill kit is designed for the sample extraction and enhanced clean-up of acrylamide in processed foods using the Waters methodology. It includes consumables for an additional mixed-mode SPE clean-up step, suitable for adoption onto mid-tier UPLC-MS/MS platforms. It provides a complete sample preparation workflow for acrylamide detection in processed foods and supports enhanced solid-phase extraction clean-up. This kit is intended to replenish consumables for the Acrylamide Starter Kit.
  • Number of analysis: 96
  • Sample preparation workflow
  • One package per kit
  • UPLC separation mode
  • MS/MS detection method
  • LC-MS system type
  • Food application
  • Application kits product type
  • One unit per package
  • Refill kit for acrylamide starter kit
  • Compatibility with mid-tier UPLC-MS/MS platforms