Learn More
Waters Corp Refill Kit, LC-MS, Acrylamide Sample Preparation, 96 Analyses
Catalog No. 502392291 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This refill kit provides necessary consumables for the sample extraction and cleanup of acrylamide in processed foods, designed for use with a specific methodology. It supports adoption on mid-tier UPLC-MS/MS platforms, like the ACQUITY I-Class Plus coupled to a XEVO TQ-S micro. The kit enables continued acrylamide analysis, particularly for educational purposes, offering high sensitivity and resolution to facilitate method adoption.
  • Contains consumables for acrylamide sample extraction and cleanup.
  • Designed for 96 analyses.
  • Supports sample preparation workflow.
  • Utilizes UPLC separation mode.
  • Employs MS/MS detection method.
  • Compatible with LC-MS system types.
  • Suitable for food applications.
  • Comes as application kits.
  • Includes 1 unit per package.
  • Offers high sensitivity and resolution for method adoption.
Catalog No. 50-239-2291 Supplier Waters Corp Supplier No. 176004418
This refill kit provides necessary consumables for the sample extraction and cleanup of acrylamide in processed foods, designed for use with a specific methodology. It supports adoption on mid-tier UPLC-MS/MS platforms, like the ACQUITY I-Class Plus coupled to a XEVO TQ-S micro. The kit enables continued acrylamide analysis, particularly for educational purposes, offering high sensitivity and resolution to facilitate method adoption.
  • Contains consumables for acrylamide sample extraction and cleanup.
  • Designed for 96 analyses.
  • Supports sample preparation workflow.
  • Utilizes UPLC separation mode.
  • Employs MS/MS detection method.
  • Compatible with LC-MS system types.
  • Suitable for food applications.
  • Comes as application kits.
  • Includes 1 unit per package.
  • Offers high sensitivity and resolution for method adoption.