Learn More
Waters Corp Acrylamide Starter Kit, LC-MS, Enhanced Cleanup
Catalog No. 502392574 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides the consumables for the extraction, enhanced cleanup, and separation of acrylamide in processed foods using a specialized methodology. It includes an additional mixed-mode SPE cleanup step for improved sample cleanup, facilitating method adoption onto mid-tier UPLC-MS/MS platforms, and is designed for ease of use, particularly for those learning about LC-MS equipment.
  • Includes an additional mixed-mode SPE cleanup step for superior sample cleanup.
  • Contains all necessary consumables and SPE equipment.
  • Features a unique chemical to improve separation sensitivity.
  • Designed for easy methodology adoption onto UPLC-MS/MS platforms.
  • For the analysis of acrylamide in processed foods.
  • Supports 96 analyses.
  • Provides a streamlined workflow for improved accuracy.
  • Aids learning for food testing with LC-MS equipment.
Catalog No. 50-239-2574 Supplier Waters Corp Supplier No. 176004419
This kit provides the consumables for the extraction, enhanced cleanup, and separation of acrylamide in processed foods using a specialized methodology. It includes an additional mixed-mode SPE cleanup step for improved sample cleanup, facilitating method adoption onto mid-tier UPLC-MS/MS platforms, and is designed for ease of use, particularly for those learning about LC-MS equipment.
  • Includes an additional mixed-mode SPE cleanup step for superior sample cleanup.
  • Contains all necessary consumables and SPE equipment.
  • Features a unique chemical to improve separation sensitivity.
  • Designed for easy methodology adoption onto UPLC-MS/MS platforms.
  • For the analysis of acrylamide in processed foods.
  • Supports 96 analyses.
  • Provides a streamlined workflow for improved accuracy.
  • Aids learning for food testing with LC-MS equipment.