Learn More
Waters Corp Acrylamide Starter Kit, LC-MS, 96 Analysis, UPLC Separation
Catalog No. 502391140 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides the consumables necessary for the sample extraction, cleanup, and separation of acrylamide in processed foods. It is designed for use with mid-tier UPLC-MS/MS platforms, such as the ACQUITY I-Class Plus coupled to a XEVO TQ-S micro. The kit facilitates the adoption of the methodology for acrylamide detection and offers everything needed for initial food testing, providing incredible resolution in separation.
  • 96 analyses
  • Sample preparation workflow
  • 1 package per kit
  • UPLC separation mode
  • MS/MS detection method
  • Refer to storage recommendations of each sub-package
  • LC-MS system type
  • Refer to shelf life recommendations of each sub-package
  • Replacement as required
  • Application in food testing
  • Application kit
  • 1 unit per package
Catalog No. 50-239-1140 Supplier Waters Corp Supplier No. 176004417
This kit provides the consumables necessary for the sample extraction, cleanup, and separation of acrylamide in processed foods. It is designed for use with mid-tier UPLC-MS/MS platforms, such as the ACQUITY I-Class Plus coupled to a XEVO TQ-S micro. The kit facilitates the adoption of the methodology for acrylamide detection and offers everything needed for initial food testing, providing incredible resolution in separation.
  • 96 analyses
  • Sample preparation workflow
  • 1 package per kit
  • UPLC separation mode
  • MS/MS detection method
  • Refer to storage recommendations of each sub-package
  • LC-MS system type
  • Refer to shelf life recommendations of each sub-package
  • Replacement as required
  • Application in food testing
  • Application kit
  • 1 unit per package