Learn More
Waters Corp Acrylamide Starter Kit, UHPLC, Enhanced Cleanup, 96 Analysis, Food Application
Catalog No. 502391679 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit offers essential consumables for the extraction, enhanced cleanup, and separation of acrylamide in processed foods. Featuring an additional mixed-mode SPE cleanup step, it improves sample quality for lower-tier U(H)PLC-MS/MS platforms, leading to cleaner samples, increased resolution, and sensitivity, especially for complex matrices.
  • 96 analyses
  • Sample preparation workflow
  • HPLC separation mode
  • MS/MS detection method
  • LC-MS system type
  • Food application
  • Enhanced cleanup for complex matrices
  • Compatible with UHPLC-MS/MS platforms
Catalog No. 50-239-1679 Supplier Waters Corp Supplier No. 176004423
This kit offers essential consumables for the extraction, enhanced cleanup, and separation of acrylamide in processed foods. Featuring an additional mixed-mode SPE cleanup step, it improves sample quality for lower-tier U(H)PLC-MS/MS platforms, leading to cleaner samples, increased resolution, and sensitivity, especially for complex matrices.
  • 96 analyses
  • Sample preparation workflow
  • HPLC separation mode
  • MS/MS detection method
  • LC-MS system type
  • Food application
  • Enhanced cleanup for complex matrices
  • Compatible with UHPLC-MS/MS platforms