Learn More
Cole-Parmer Argos Technologies ACP05 CleanCut™ Acrylic Paper Towel Dispenser 10.5 x 4 x 6.38"; 1/EA
Catalog No. 501960911 Shop All Cole-Parmer Products
Encompass_Preferred
Argos Technologies CleanCut acrylics is a range of clean line storage solutions perfect for laboratory organization. Made of clear durable acrylic, these holders offer versatility while providing space-saving storage. All mounting accessories included. Paper Towel Holder: Simply designed to accommodate a variety of C-fold or Z-fold paper towels. Can be wall mounted or placed directly on the benchtop. Glove Box Holders: Heat-bent thick walls simplify construction and limit accidental breakage. Triple and quad holders have a unique design that allows glove boxes to be loaded from either the top or side. Can be mounted vertically or horizontally. standard glove boxes accommodating depths up to 3.75". Safety Glasses Holder: Built-in handles on either side allow for easy setup. Holder can be mounted vertically, horizontally or placed on the benchtop.
Catalog No. 50-196-0911 Supplier Cole-Parmer Supplier No. 6353525
Argos Technologies CleanCut acrylics is a range of clean line storage solutions perfect for laboratory organization. Made of clear durable acrylic, these holders offer versatility while providing space-saving storage. All mounting accessories included. Paper Towel Holder: Simply designed to accommodate a variety of C-fold or Z-fold paper towels. Can be wall mounted or placed directly on the benchtop. Glove Box Holders: Heat-bent thick walls simplify construction and limit accidental breakage. Triple and quad holders have a unique design that allows glove boxes to be loaded from either the top or side. Can be mounted vertically or horizontally. standard glove boxes accommodating depths up to 3.75". Safety Glasses Holder: Built-in handles on either side allow for easy setup. Holder can be mounted vertically, horizontally or placed on the benchtop.