Learn More
Research Products International Corp Acrylic Compartment Box, 15 Compartments, 13 5/8 x 6 1/2, x 2 Inches
SDP
Catalog No. 50212339
Encompass
Unique Acrylic Compartment Boxes are specially designed for use in freezers where temperatures reach -70°C. Compartment boxes are ideal for storing various size bottles, vials, enzyme reagents, specimens, etc. Boxes organize contents and allow inventory control and convenient stock rotation. Constructed of 1/8 inch thick transparent acrylic with a hinged lid. Up to six specially designed racks for 1.5ml Micro-Centrifuge Tubes fit neatly inside the compartment box. Racks hold twenty 1.5 - 2.0 ml micro-tubes.
Catalog No. 50-212-339 Supplier Research Products International Corp Supplier No. 145736
Unique Acrylic Compartment Boxes are specially designed for use in freezers where temperatures reach -70°C. Compartment boxes are ideal for storing various size bottles, vials, enzyme reagents, specimens, etc. Boxes organize contents and allow inventory control and convenient stock rotation. Constructed of 1/8 inch thick transparent acrylic with a hinged lid. Up to six specially designed racks for 1.5ml Micro-Centrifuge Tubes fit neatly inside the compartment box. Racks hold twenty 1.5 - 2.0 ml micro-tubes.