Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acrylic Acid N-Hydroxysuccinimide Ester

Catalog No. A80601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A80601G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. A80601G Supplier Sigma Aldrich Supplier No. A80601G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acrylic acid N-hydroxysuccinimide ester (NAS) is a general acrylating agent that is used in the synthesis of:Acrylated hyaluronic acid (HA) hydrogel for bone regeneration applications.Injectable, biodegradable, and thermosensitive copolymers for tissue engineering.Imidazole-based biocompatible, aqueous quantum dots (QDs).

Specifications

CAS 295341-43-4
Melting Point 236°C to 241°C
Molecular Formula C2H5NaO2S
Linear Formula C2H5NaO2S
Quantity 500 mg
Molecular Weight (g/mol) 116.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.