Learn More
Braintree Scientific Acrylic Adjustable Tissue Slicer,Curved 12mm D up to 25mm W
SDP
Catalog No. 501956055
Encompass
Adjustable All-Purpose Acrylic Channel or Cavity Tissue/Brain Matrice. Acrylic all-purpose tissue/brain matrice has an adjustable cutting width and can be used to generate 1mm incremental thick sections in all types of tissues. The adjustable length of cutting surface is from 1mm to 25mm. The flexibility of these acrylic matrices makes it ideal for many different tissue-slicing functions. Order S for a sagittal style, and C for a coronal style. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6055 Supplier Braintree Scientific Supplier No. TMC12
Adjustable All-Purpose Acrylic Channel or Cavity Tissue/Brain Matrice. Acrylic all-purpose tissue/brain matrice has an adjustable cutting width and can be used to generate 1mm incremental thick sections in all types of tissues. The adjustable length of cutting surface is from 1mm to 25mm. The flexibility of these acrylic matrices makes it ideal for many different tissue-slicing functions. Order S for a sagittal style, and C for a coronal style. **Sold for research purposes only and not for human use**