Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acrylic anhydride

Catalog No. 9092701G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-927-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-927-01G Supplier Sigma Aldrich Supplier No. 9092701G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acrylic anhydride can be used to prepare specialty acrylate, acrylamide monomers or acrylic resin. It forms cyclic anhydrides on polymerization and can not be used as a crosslinker.

Specifications

CAS 2051-76-5
Molecular Formula C6H6O3
Linear Formula C6H6O3
Quantity 1 g
MDL Number MFCD00048146
Molecular Weight (g/mol) 126.11
Percent Purity ≥90%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.