Learn More
World Precision Instrument Acrylic Brain Matrix for Rat, 300g-600g, Coronal Slices
SDP
Catalog No. 502538602
Encompass
Using a WPI brain matrix, you can get thin and uniform brain slices, which is crucial for histological analysis, immunohistochemistry, molecular biology, and more. This consistency is essential for accurate comparisons between different brains and experimental conditions. These WPI rodent brain matrices are made of acrylic, a durable material. They have precision-cut channels or slots that hold the brain in place during slicing. Choose your brain matrix based on your species (mouse or rat) and the size of the animal. The WPI brain matrices are designed for sagittal or coronal slices. These brain matrices give you consistent slices that are 1 mm thick, and the olfactory spinal notch is cut into every brain matrix. Get standardized and precise brain sectioning. Use WPI brain matrices to ensure the reproducibility and accuracy of your experiments for a better understanding of the structure and function of the rodent brain.
Catalog No. 50-253-8602 Supplier World Precision Instrument Supplier No. RBMA600C
Using a WPI brain matrix, you can get thin and uniform brain slices, which is crucial for histological analysis, immunohistochemistry, molecular biology, and more. This consistency is essential for accurate comparisons between different brains and experimental conditions. These WPI rodent brain matrices are made of acrylic, a durable material. They have precision-cut channels or slots that hold the brain in place during slicing. Choose your brain matrix based on your species (mouse or rat) and the size of the animal. The WPI brain matrices are designed for sagittal or coronal slices. These brain matrices give you consistent slices that are 1 mm thick, and the olfactory spinal notch is cut into every brain matrix. Get standardized and precise brain sectioning. Use WPI brain matrices to ensure the reproducibility and accuracy of your experiments for a better understanding of the structure and function of the rodent brain.