Learn More
Electron Microscopy Sciences Acrylic Desiccator Cabinet, 10" x 7", 8-1/2" height
SDP
Catalog No. 50930910
Encompass_Preferred
For dry, dust-free storage at temperatures up to 120 degree F. They can be used as incubators, freezers, and refrigerators or at room temperature. Cabinets have airtight doors that need no lubrication. Sliding perforated shelves for easy access; transparent acrylic walls for see-throguh storage. The third bottom shelf accommodates desiccants. Cabinets are also available with locks and a hygrometer.
Hygrometer has a temperature range of -58 degree F to 158 degree F with +/-1.5 degree F accuracy up to 104 degree F. Humidity range is 20 to 95% with +/-5% accuracy. LCD display. Includes 2 'AAA' batteries.
Footprint: 10" x 7"
Height: 8-1/2"
Catalog No. 50-930-910 Supplier Electron Microscopy Sciences Supplier No. 7122010
For dry, dust-free storage at temperatures up to 120 degree F. They can be used as incubators, freezers, and refrigerators or at room temperature. Cabinets have airtight doors that need no lubrication. Sliding perforated shelves for easy access; transparent acrylic walls for see-throguh storage. The third bottom shelf accommodates desiccants. Cabinets are also available with locks and a hygrometer.
Hygrometer has a temperature range of -58 degree F to 158 degree F with +/-1.5 degree F accuracy up to 104 degree F. Humidity range is 20 to 95% with +/-5% accuracy. LCD display. Includes 2 'AAA' batteries.
Footprint: 10" x 7"
Height: 8-1/2"