Learn More
Cole-Parmer Plas-Labs 860-CG Acrylic Desiccator with Gas Port, Clear; 12"W x 12"H x 12"D
Catalog No. NC0949317 Shop All Cole-Parmer Products
Encompass
These durable desiccators have a temperature range of 40 to 120F (4 to 49C). They are made of heavy-duty, 1/4" thick acrylic. Closed cellular PVC gaskets prevent system leaksgaskets will not take a set or keep an impression. Two gas ports (1/8" NPT x 1/4" hose barb inlet; 1/4" NPT x 1/4" hose barb outlet) are included. Not designed for vacuum use. Choose clear units for easy viewing of contents, or choose amber units for protecting light-sensitive compounds from ultraviolet light. All models are easy to access and clean; slip-away hinges allow complete removal of door when needed. Adjust shelf positions to suit your needs. Models 08900-50 and -52 include two shelves; will hold five shelves. All other models include four shelves; will hold ten shelves.
Catalog No. NC0949317 Supplier Cole-Parmer Supplier No. 0890050
These durable desiccators have a temperature range of 40 to 120F (4 to 49C). They are made of heavy-duty, 1/4" thick acrylic. Closed cellular PVC gaskets prevent system leaksgaskets will not take a set or keep an impression. Two gas ports (1/8" NPT x 1/4" hose barb inlet; 1/4" NPT x 1/4" hose barb outlet) are included. Not designed for vacuum use. Choose clear units for easy viewing of contents, or choose amber units for protecting light-sensitive compounds from ultraviolet light. All models are easy to access and clean; slip-away hinges allow complete removal of door when needed. Adjust shelf positions to suit your needs. Models 08900-50 and -52 include two shelves; will hold five shelves. All other models include four shelves; will hold ten shelves.