Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Variable-Area Flowmeter Kit without Valve, Direct Reading, Acrylic Housing and Stainless Steel Fittings; 8.5 L/min Air
Catalog No. NC1017427
Encompass
These easy-to-read acrylic flowmeter kits come with seven interchangeable back plates for routine gases air, argon, nitrogen, helium, carbon dioxide, oxygen, and water. Meters are offered with or without built-in low hysteresis needle valves. Meters without valves can be panel mounted or used in partial or full in-line mounted configurations.
Catalog No. NC1017427 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3450040
These easy-to-read acrylic flowmeter kits come with seven interchangeable back plates for routine gases air, argon, nitrogen, helium, carbon dioxide, oxygen, and water. Meters are offered with or without built-in low hysteresis needle valves. Meters without valves can be panel mounted or used in partial or full in-line mounted configurations.