Learn More
MSC Misco Products Corporation FIRST STEP Acrylic Floor Sealer, 5 Gallon, 18% Non-Volatile Solids
Catalog No. 502838027
Encompass
An acrylic floor sealer formulated to provide a hard, durable undercoat for use prior to the application of any floor finish. It never yellows and will continue to protect floors after all floor finish has worn off. It provides chemical and stain resistance and is suitable for use on any type of hard surface.
  • Acrylic based sealer
  • Compatible
  • Versatile
  • Protects flooring
  • Easy to apply
  • Use prior to any acrylic floor finish
  • Use on any type of hard surface
  • Resists chemicals and stains
Catalog No. 50-283-8027 Supplier MSC Supplier No. 63348379
An acrylic floor sealer formulated to provide a hard, durable undercoat for use prior to the application of any floor finish. It never yellows and will continue to protect floors after all floor finish has worn off. It provides chemical and stain resistance and is suitable for use on any type of hard surface.
  • Acrylic based sealer
  • Compatible
  • Versatile
  • Protects flooring
  • Easy to apply
  • Use prior to any acrylic floor finish
  • Use on any type of hard surface
  • Resists chemicals and stains