Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Acrylic Glove Box Holder - Wall Mount
SDP

Catalog No. 01259214
Change view
Click to view available options
Capacity:
1 Box
2 Boxes
3 Boxes
4 Boxes
5 Boxes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
01-259-214 3 Boxes
01-259-184 1 Box
01-259-149 2 Boxes
01-259-209 4 Boxes
01-259-186 5 Boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
N/A
Catalog No. 01-259-214 Supplier Poltex Supplier No. GBACOV3W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N/A

Acrylic glove box holder available in 1, 2, 3, 4, or 5 box capacity. Mount vertically or horizontally with wall mount hardware and durable 1/4 in. acrylic construction.

  • Acrylic glove box holder
  • Available in 1, 2, 3, 4, or 5 box capacity
  • Can be mounted vertically or horizontally
  • Wall mount (screws and anchors included)
  • Durable 1/4 in. acrylic construction
  • Consumables not included

Specifications

Product Type Glove Box Holder
Color Clear
Dimensions (L x W x H) 4 x 17.8 x 9.3 in.
Holds Exam Glove Boxes, Disposable Glove Boxes, Sterile Gloves, Glove and Tssue Boxes, Gloves
Material PETG
Mounting Wall Mount
No. of Compartments 3
Product Style Side Loading
Capacity 3 Boxes
Edges Rounded
Formulation PETG
For Use With (Application) Glove Box Holder
Includes Wall Mount Hardware
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.