Learn More
Ted Pella Inc Acrylic Brain Matrices, Mouse, 30g Adult, Coronal, 1mm
SDP
Catalog No. NC0888017
Encompass
An economical alternative to metal brain matrices. The acrylic coronal brain matrices have the additional feature of a mid-line sagittal cut to facilitate the separation of the left and right hemispheres. All divisions are 1mm apart.
Catalog No. NC0888017 Supplier Ted Pella Inc Supplier No. 15050
Add to Cart
Add to Cart
An economical alternative to metal brain matrices. The acrylic coronal brain matrices have the additional feature of a mid-line sagittal cut to facilitate the separation of the left and right hemispheres. All divisions are 1mm apart.