Learn More
Braintree Scientific Acrylic Mouse Matrice - 1mm sagittal
SDP
Catalog No. 501954385
Encompass
Acrylic Matrices - High grade acrylic matrices are an economical method to obtain even slices of brain tissues from rats and mice. These units include the spinal and olfactory bulb notches. Order S for a sagittal style, and C for a coronal style. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4385 Supplier Braintree Scientific Supplier No. BSA5000S
Acrylic Matrices - High grade acrylic matrices are an economical method to obtain even slices of brain tissues from rats and mice. These units include the spinal and olfactory bulb notches. Order S for a sagittal style, and C for a coronal style. **Sold for research purposes only and not for human use**