Learn More
Research Products International Corp Acrylic Organizer for Styrofoam Storage Boxes, 2 x 2 Array, 12 1/2 x 8 1/2 x 7 Inches
SDP
Catalog No. 50212835
Encompass
Acrylic Organizers are convenient for storing and organizing styrofoam storage boxes. Constructed of 1/8 inch thick acrylic for durability and clarity. Perfect for use in a refrigerator, freezer or on a bench top.
Catalog No. 50-212-835 Supplier Research Products International Corp Supplier No. 145759B
Acrylic Organizers are convenient for storing and organizing styrofoam storage boxes. Constructed of 1/8 inch thick acrylic for durability and clarity. Perfect for use in a refrigerator, freezer or on a bench top.