Learn More
Electron Microscopy Sciences Acrylic Plastic Casting 1 QT
SDP
Catalog No. 5031096
Encompass_Preferred
Simply place your object in a mold, mix and pour Acrylic Plastic over it. In an hour it is hard and your object is firmly embedded in a clear, stable plastic. For high-gloss finish, it can be polished with a soft cloth. Mold making and casting can be done in two hours. Acrylic Plastic Casting is a thermosetting acrylic-polymer plastic with excellent clarity and stabile characteristics. The kit is available with 1 pint of resin and hardener, or 1quart of resin and hardener.
Catalog No. 50-310-96 Supplier Electron Microscopy Sciences Supplier No. 2421002
Simply place your object in a mold, mix and pour Acrylic Plastic over it. In an hour it is hard and your object is firmly embedded in a clear, stable plastic. For high-gloss finish, it can be polished with a soft cloth. Mold making and casting can be done in two hours. Acrylic Plastic Casting is a thermosetting acrylic-polymer plastic with excellent clarity and stabile characteristics. The kit is available with 1 pint of resin and hardener, or 1quart of resin and hardener.