Learn More
Supertek™ Acrylic Prism
GREENER_CHOICESCI_ED
Catalog No. S01250 Shop All Supertek Scientific Products
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S01250 Acrylic
1 options
Catalog No. S01250 Supplier Supertek™ Supplier No. PH30305
Add to Cart
Add to Cart
This Acrylic Prism Set consists of crystal-clear acrylic equilateral prisms.
  • Acrylic Prism Set of 3 consists of Crystal-clear acrylic equilateral prisms 25 mm faces, 1 25 mm long, 1 50mm long and 1 100mm long.
Specifications
Material Acrylic
Product Type Equilateral Prism