Learn More
Braintree Scientific Acrylic Rat Matrice - 1mm Coronal (300-600g)
SDP
Catalog No. 501954388
Encompass
Acrylic Matrices - High grade acrylic matrices are an economical method to obtain even slices of brain tissues from rats and mice. These units include the spinal and olfactory bulb notches. Order S for a sagittal style, and C for a coronal style. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4388 Supplier Braintree Scientific Supplier No. BSA7000C
Acrylic Matrices - High grade acrylic matrices are an economical method to obtain even slices of brain tissues from rats and mice. These units include the spinal and olfactory bulb notches. Order S for a sagittal style, and C for a coronal style. **Sold for research purposes only and not for human use**