Learn More
US Plastic Corporation 0.125" (3.2mm) x 12" x 12" White 7328 Acrylic Sheet
SDP
Catalog No. NC0724512
Encompass
Acrylic sheets are lightweight and have many applications where weather and impact resistance is important. This material is excellent to use in place of glass, where breakage is a threat to safety. Sheets are UV stabilized. White is translucent and provides a low level of light transmission. White #7328 allows 31% of light transmit. Forming temperature is 290°F to 320°F.
Catalog No. NC0724512 Supplier US Plastic Corporation Supplier No. 44440
Acrylic sheets are lightweight and have many applications where weather and impact resistance is important. This material is excellent to use in place of glass, where breakage is a threat to safety. Sheets are UV stabilized. White is translucent and provides a low level of light transmission. White #7328 allows 31% of light transmit. Forming temperature is 290°F to 320°F.