Learn More
World Precision Instrument Acrylic Surgical Table
SDP
Catalog No. 502521383
Encompass
Constructed of clear acrylic, this economical surgery table features adjustable nylon cords for fast animal restraint and a slight incline for easy viewing. The acrylic construction is durable, yet easy to clean. The nylon cords may be adjusted at the point of contact with the animal and also under the table itself. The surgery table is designed for use in dissecting small animals such as frogs, birds, mice, rats, guinea pigs, etc.
Catalog No. 50-252-1383 Supplier World Precision Instrument Supplier No. 501892
Constructed of clear acrylic, this economical surgery table features adjustable nylon cords for fast animal restraint and a slight incline for easy viewing. The acrylic construction is durable, yet easy to clean. The nylon cords may be adjusted at the point of contact with the animal and also under the table itself. The surgery table is designed for use in dissecting small animals such as frogs, birds, mice, rats, guinea pigs, etc.