Learn More
MSC Tape Logic Acrylic Tape, Clear, 2 in, 36 Rolls
Catalog No. 501919307
Encompass
Tape Logic #400 Industrial Tape is an acrylic tape best suited for long-term storage where shelf life is important. This clear acrylic tape maintains its clarity over extended periods and will not yellow with age. It offers a consistent, smooth release and a strong adhesive that grips well to recycled corrugated materials.
  • Acrylic tape is best for long term storage when shelf life is important.
  • Clear acrylic tape retains its clarity over long periods of time.
  • Will not yellow over time.
  • Consistent, smooth release.
  • Strong adhesive grips well to recycled corrugated.
  • Application temperature range 32° - 140° F.
  • Meets UPS and U.S. postal regulations.
Catalog No. 50-191-9307 Supplier MSC Supplier No. 39589205
Tape Logic #400 Industrial Tape is an acrylic tape best suited for long-term storage where shelf life is important. This clear acrylic tape maintains its clarity over extended periods and will not yellow with age. It offers a consistent, smooth release and a strong adhesive that grips well to recycled corrugated materials.
  • Acrylic tape is best for long term storage when shelf life is important.
  • Clear acrylic tape retains its clarity over long periods of time.
  • Will not yellow over time.
  • Consistent, smooth release.
  • Strong adhesive grips well to recycled corrugated.
  • Application temperature range 32° - 140° F.
  • Meets UPS and U.S. postal regulations.