Learn More
MSC 2X55Yd Tan Acrylic Tape
Catalog No. 501909941
Encompass
2" x 55 Yd Tan Acrylic Adhesive Packaging Tape Polypropylene Film Backing, 1.8 mil Thick, 18 Lb Tensile Strength
Catalog No. 50-190-9941 Supplier MSC Supplier No. 39478433
2" x 55 Yd Tan Acrylic Adhesive Packaging Tape Polypropylene Film Backing, 1.8 mil Thick, 18 Lb Tensile Strength