Learn More
MSC Tape Logic Packing Tape, 3 in Wide, Clear, 6 Rolls
Catalog No. 501918543
Encompass
Industrial Carton Sealing Tape with a consistent smooth release. This tape is ideal for long-term storage, maintaining clarity and resisting yellowing, and features a strong acrylic adhesive for reliable grip on recycled corrugated boxes.
  • Consistent smooth release
  • Retains clarity and will not yellow for long-term storage
  • Strong adhesive grips well to recycled corrugated boxes
  • Application temperature range of 32-140 degrees Fahrenheit
  • Meets UPS and US Postal regulations
  • Best for lightweight carton sealing applications
  • Polypropylene material
  • Has 2 mil thickness
  • Achieves 20 lb/inch tensile strength
Catalog No. 50-191-8543 Supplier MSC Supplier No. 39581335
Industrial Carton Sealing Tape with a consistent smooth release. This tape is ideal for long-term storage, maintaining clarity and resisting yellowing, and features a strong acrylic adhesive for reliable grip on recycled corrugated boxes.
  • Consistent smooth release
  • Retains clarity and will not yellow for long-term storage
  • Strong adhesive grips well to recycled corrugated boxes
  • Application temperature range of 32-140 degrees Fahrenheit
  • Meets UPS and US Postal regulations
  • Best for lightweight carton sealing applications
  • Polypropylene material
  • Has 2 mil thickness
  • Achieves 20 lb/inch tensile strength