Learn More
Bio Rad Laboratories ACRYLMIDE BIS SOLUTION 30%
Catalog No. NC9376992
Encompass
Electrophoresis Gel Component; 30% Acrylamide/Bis Solution, 29:1; Bio-Rad; 500mL; Ready to use; Made from 99.9% pure acrylamide and bis-acrylamide
Catalog No. NC9376992 Supplier Bio Rad Laboratories Supplier No. 1610156
Electrophoresis Gel Component; 30% Acrylamide/Bis Solution, 29:1; Bio-Rad; 500mL; Ready to use; Made from 99.9% pure acrylamide and bis-acrylamide